gRNA Design Details

Target DNA Sequence
ACGCGCCATTGGAATGTCCC
PAM Sequence
TGG
Target locus
Chr.9: 91220951 - 91220973 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 90.52/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
AUH
Entrez Gene ID
549
Gene Name
AU RNA binding protein/enoyl-CoA hydratase
Gene Aliases
-
Chromosome Location
Chr.9: 91213815 - 91361969 on GRCh38
UniGene ID
Hs.175905
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001306190.1 NP_001293119.1 6
NM_001698.2 NP_001689.1 7
XM_005252066.3 XP_005252123.1 8
XM_005252067.4 XP_005252124.1 8
XM_005252069.4 XP_005252126.1 8
XM_006717150.3 XP_006717213.1 7
XM_011518802.2 XP_011517104.1 8
XM_017014849.1 XP_016870338.1 7
XM_017014850.1 XP_016870339.1 8
XM_017014851.1 XP_016870340.1 8