gRNA Design Details
- Target DNA Sequence
- GCTGCTTTAACGTGATCAAT
- PAM Sequence
- AGG
- Target locus
-
Chr.17: 34320293 - 34320315 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
89.9/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CCL8
- Entrez Gene ID
-
6355
- Gene Name
- C-C motif chemokine ligand 8
- Gene Aliases
-
HC14, MCP-2, MCP2, SCYA10, SCYA8
- Chromosome Location
-
Chr.17: 34319047 - 34321402 on GRCh38
- UniGene ID
- Hs.271387
- Species
-
Human