gRNA Design Details
- Target DNA Sequence
- GCGTGAGCTATTACATGGAG
- PAM Sequence
- GGG
- Target locus
-
Chr.22: 46536313 - 46536335 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
98.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CELSR1
- Entrez Gene ID
-
9620
- Gene Name
- cadherin EGF LAG seven-pass G-type receptor 1
- Gene Aliases
-
ADGRC1, CDHF9, FMI2, HFMI2, ME2
- Chromosome Location
-
Chr.22: 46360834 - 46537502 on GRCh38
- UniGene ID
- Hs.252387
- Species
-
Human