gRNA Design Details
- Target DNA Sequence
- AGTCGATTCGGAAGTAGCCC
- PAM Sequence
- CGG
- Target locus
-
Chr.22: 46536273 - 46536295 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
96.18/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CELSR1
- Entrez Gene ID
-
9620
- Gene Name
- cadherin EGF LAG seven-pass G-type receptor 1
- Gene Aliases
-
ADGRC1, CDHF9, FMI2, HFMI2, ME2
- Chromosome Location
-
Chr.22: 46360834 - 46537502 on GRCh38
- UniGene ID
- Hs.252387
- Species
-
Human