gRNA Design Details
- Target DNA Sequence
- ACGTTCGCCACCTTCTGGAT
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 207768021 - 207768043 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
94.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FZD5
- Entrez Gene ID
-
7855
- Gene Name
- frizzled class receptor 5
- Gene Aliases
-
C2orf31, HFZ5
- Chromosome Location
-
Chr.2: 207762586 - 207769419 on GRCh38
- UniGene ID
- Hs.17631
- Species
-
Human