gRNA Design Details

Target DNA Sequence
AGACGGCCATTCAGCTTATG
PAM Sequence
GGG
Target locus
Chr.1: 71046720 - 71046742 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 96.18/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PTGER3
Entrez Gene ID
5733
Gene Name
prostaglandin E receptor 3
Gene Aliases
EP3, EP3-I, EP3-II, EP3-III, EP3-IV, EP3-VI, EP3e, PGE2-R
Chromosome Location
Chr.1: 70852353 - 71047814 on GRCh38
UniGene ID
Hs.445000
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001126044.1 NP_001119516.1 1
NM_198714.1 NP_942007.1 1
NM_198715.2 NP_942008.1 1
NM_198716.1 NP_942009.1 1
NM_198717.1 NP_942010.1 1
NM_198718.1 NP_942011.1 1
NM_198719.1 NP_942012.1 1
NR_028292.1 1
NR_028293.1 1
NR_028294.1 1
XM_011541810.2 XP_011540112.1 1