gRNA Design Details

Target DNA Sequence
GAGATTCTGGAACGCGTAGT
PAM Sequence
CGG
Target locus
Chr.1: 202276482 - 202276504 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 91.41/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
LGR6
Entrez Gene ID
59352
Gene Name
leucine rich repeat containing G protein-coupled receptor 6
Gene Aliases
GPCR, VTS20631
Chromosome Location
Chr.1: 202193990 - 202319761 on GRCh38
UniGene ID
Hs.497402
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001017403.1 NP_001017403.1 5
NM_021636.2 NP_067649.2 5
XM_005245404.3 XP_005245461.1 5
XM_011509838.2 XP_011508140.1 5
XM_011509839.2 XP_011508141.1 5
XM_011509841.2 XP_011508143.1 2
XM_011509842.2 XP_011508144.1 2
XM_011509843.2 XP_011508145.1 2
XM_011509846.2 XP_011508148.1 2
XM_017001996.1 XP_016857485.1 5
XM_017001997.1 XP_016857486.1 3