gRNA Design Details
- Target DNA Sequence
- GACGACGAGTACGGCCGGCA
- PAM Sequence
- GGG
- Target locus
-
Chr.1: 1332174 - 1332196 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
94.27/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TAS1R3
- Entrez Gene ID
-
83756
- Gene Name
- taste 1 receptor member 3
- Gene Aliases
-
T1R3
- Chromosome Location
-
Chr.1: 1331324 - 1335320 on GRCh38
- UniGene ID
- Hs.74375
- Species
-
Human