gRNA Design Details
- Target DNA Sequence
- AACCTCGGGGAGTGCCACTT
- PAM Sequence
- CGG
- Target locus
-
Chr.3: 114131155 - 114131177 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
91.96/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- DRD3
- Entrez Gene ID
-
1814
- Gene Name
- dopamine receptor D3
- Gene Aliases
-
D3DR, ETM1, FET1
- Chromosome Location
-
Chr.3: 114127623 - 114199407 on GRCh38
- UniGene ID
- Hs.121478
- Species
-
Human