gRNA Design Details

Target DNA Sequence
TACGTGGACAAGCGATTGGC
PAM Sequence
GGG
Target locus
Chr.5: 40681381 - 40681403 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 94.82/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PTGER4
Entrez Gene ID
5734
Gene Name
prostaglandin E receptor 4
Gene Aliases
EP4, EP4R
Chromosome Location
Chr.5: 40679930 - 40740932 on GRCh38
UniGene ID
Hs.199248
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000958.2 NP_000949.1 2
XM_017009656.1 XP_016865145.1 1
XM_017009657.1 XP_016865146.1 1
XM_017009658.1 XP_016865147.1 1
XM_017009659.1 XP_016865148.1 1