gRNA Design Details
- Target DNA Sequence
- GGAAGCCCAGCGACACGCAC
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 207767884 - 207767906 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
93.45/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FZD5
- Entrez Gene ID
-
7855
- Gene Name
- frizzled class receptor 5
- Gene Aliases
-
C2orf31, HFZ5
- Chromosome Location
-
Chr.2: 207762586 - 207769419 on GRCh38
- UniGene ID
- Hs.17631
- Species
-
Human