gRNA Design Details

Target DNA Sequence
AACAACGACCGTGGCACCGA
PAM Sequence
AGG
Target locus
Chr.22: 46433452 - 46433474 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 94.82/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CELSR1
Entrez Gene ID
9620
Gene Name
cadherin EGF LAG seven-pass G-type receptor 1
Gene Aliases
ADGRC1, CDHF9, FMI2, HFMI2, ME2
Chromosome Location
Chr.22: 46360834 - 46537502 on GRCh38
UniGene ID
Hs.252387
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_014246.1 NP_055061.1 5
XM_006724383.3 XP_006724446.1 5
XM_011530553.1 XP_011528855.1 5
XM_011530554.2 XP_011528856.1 5
XM_011530555.2 XP_011528857.1 5