gRNA Design Details
- Target DNA Sequence
- GACGTTGGCAAAGATGAAGT
- PAM Sequence
- CGG
- Target locus
-
Chr.2: 136115397 - 136115419 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
89.84/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CXCR4
- Entrez Gene ID
-
7852
- Gene Name
- C-X-C motif chemokine receptor 4
- Gene Aliases
-
CD184, D2S201E, FB22, HM89, HSY3RR, LAP-3, LAP3, LCR1, LESTR, NPY3R, NPYR, NPYRL, NPYY3R, WHIM, WHIMS
- Chromosome Location
-
Chr.2: 136114349 - 136118155 on GRCh38
- UniGene ID
- Hs.593413
- Species
-
Human