gRNA Design Details

Target DNA Sequence
GCGTTACTACAGCATCTGCC
PAM Sequence
AGG
Target locus
Chr.3: 114131343 - 114131365 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 91.7/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
DRD3
Entrez Gene ID
1814
Gene Name
dopamine receptor D3
Gene Aliases
D3DR, ETM1, FET1
Chromosome Location
Chr.3: 114127623 - 114199407 on GRCh38
UniGene ID
Hs.121478
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000796.5 NP_000787.2 6
NM_001282563.2 NP_001269492.1 7
NM_001290809.1 NP_001277738.1 7
NM_033663.5 NP_387512.3 6
XM_017005829.1 XP_016861318.1 7