gRNA Design Details

Target DNA Sequence
CAAAGCGGCCCGTGTCCGTG
PAM Sequence
AGG
Target locus
Chr.1: 11748486 - 11748508 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 92.64/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
AGTRAP
Entrez Gene ID
57085
Gene Name
angiotensin II receptor associated protein
Gene Aliases
ATRAP
Chromosome Location
Chr.1: 11736085 - 11750771 on GRCh38
UniGene ID
Hs.464438
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001040194.1 NP_001035284.1 4
NM_001040195.1 NP_001035285.1 3
NM_001040196.1 NP_001035286.1 5
NM_001040197.1 NP_001035287.1 5
NM_020350.4 NP_065083.3 4
XM_011541799.2 XP_011540101.2 5
XM_011541800.2 XP_011540102.1 5
XM_011541802.1 XP_011540104.1 6