gRNA Design Details
- Target DNA Sequence
- TCGGCCTCCACCTTCTTTGG
- PAM Sequence
- CGG
- Target locus
-
Chr.19: 14473488 - 14473510 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
91.1/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PTGER1
- Entrez Gene ID
-
5731
- Gene Name
- prostaglandin E receptor 1
- Gene Aliases
-
EP1
- Chromosome Location
-
Chr.19: 14472466 - 14475362 on GRCh38
- UniGene ID
- Hs.159360
- Species
-
Human