gRNA Design Details
- Target DNA Sequence
- GATCCCGAATGGCATAACCA
- PAM Sequence
- AGG
- Target locus
-
Chr.6: 116806420 - 116806442 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
93.38/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GPRC6A
- Entrez Gene ID
-
222545
- Gene Name
- G protein-coupled receptor class C group 6 member A
- Gene Aliases
-
GPCR, bA86F4.3
- Chromosome Location
-
Chr.6: 116788512 - 116831128 on GRCh38
- UniGene ID
- Hs.266745
- Species
-
Human