gRNA Design Details

Target DNA Sequence
CCGCTACGTGGCCATCGTCC
PAM Sequence
AGG
Target locus
Chr.17: 40555398 - 40555420 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 91.55/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CCR7
Entrez Gene ID
1236
Gene Name
C-C motif chemokine receptor 7
Gene Aliases
BLR2, CC-CKR-7, CCR-7, CD197, CDw197, CMKBR7, EBI1
Chromosome Location
Chr.17: 40553769 - 40565484 on GRCh38
UniGene ID
Hs.370036
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001301714.1 NP_001288643.1 2
NM_001301716.1 NP_001288645.1 3
NM_001301717.1 NP_001288646.1 3
NM_001301718.1 NP_001288647.1 3
NM_001838.3 NP_001829.1 3