gRNA Design Details
- Target DNA Sequence
- AGTCTGACCAAGCTCGTCCG
- PAM Sequence
- GGG
- Target locus
-
Chr.12: 12908601 - 12908623 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
94.55/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GPRC5A
- Entrez Gene ID
-
9052
- Gene Name
- G protein-coupled receptor class C group 5 member A
- Gene Aliases
-
GPCR5A, PEIG-1, RAI3, RAIG1, TIG1
- Chromosome Location
-
Chr.12: 12891022 - 12913666 on GRCh38
- UniGene ID
- Hs.631733
- Species
-
Human