gRNA Design Details
- Target DNA Sequence
- AGTTCGTTCGTCATGTTTCG
- PAM Sequence
- CGG
- Target locus
-
Chr.3: 48656176 - 48656198 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
97.27/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CELSR3
- Entrez Gene ID
-
1951
- Gene Name
- cadherin EGF LAG seven-pass G-type receptor 3
- Gene Aliases
-
ADGRC3, CDHF11, EGFL1, FMI1, HFMI1, MEGF2, RESDA1
- Chromosome Location
-
Chr.3: 48636463 - 48662915 on GRCh38
- UniGene ID
- Hs.631926
- Species
-
Human