gRNA Design Details
- Target DNA Sequence
- GTCGAGTGAGACCGGCCGTG
- PAM Sequence
- GGG
- Target locus
-
Chr.2: 48755512 - 48755534 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
88.78/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- LHCGR
- Entrez Gene ID
-
3973
- Gene Name
- luteinizing hormone/choriogonadotropin receptor
- Gene Aliases
-
HHG, LCGR, LGR2, LH/CG-R, LH/CGR, LHR, LHRHR, LSH-R, ULG5
- Chromosome Location
-
Chr.2: 48686774 - 48755741 on GRCh38
- UniGene ID
- Hs.468490
- Species
-
Human