gRNA Design Details
- Target DNA Sequence
- CTTCGATCCGTAGGGTGTAA
- PAM Sequence
- GGG
- Target locus
-
Chr.4: 61733232 - 61733254 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
95.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRL3
- Entrez Gene ID
-
23284
- Gene Name
- adhesion G protein-coupled receptor L3
- Gene Aliases
-
CIRL3, CL3, LEC3, LPHN3
- Chromosome Location
-
Chr.4: 61201256 - 62078335 on GRCh38
- UniGene ID
- Hs.28391
- Species
-
Human