gRNA Design Details
- Target DNA Sequence
- GCTTCATCAATCGCAACTAC
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 236581412 - 236581434 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
91.85/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ACKR3
- Entrez Gene ID
-
57007
- Gene Name
- atypical chemokine receptor 3
- Gene Aliases
-
CMKOR1, CXC-R7, CXCR-7, CXCR7, GPR159, RDC-1, RDC1
- Chromosome Location
-
Chr.2: 236537131 - 236582358 on GRCh38
- UniGene ID
- Hs.471751
- Species
-
Human