gRNA Design Details

Target DNA Sequence
GAACTACATGCGCTGCGTGT
PAM Sequence
CGG
Target locus
Chr.1: 109258835 - 109258857 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 95.04/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CELSR2
Entrez Gene ID
1952
Gene Name
cadherin EGF LAG seven-pass G-type receptor 2
Gene Aliases
ADGRC2, CDHF10, EGFL2, Flamingo1, MEGF3
Chromosome Location
Chr.1: 109249705 - 109275756 on GRCh38
UniGene ID
Hs.57652
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001408.2 NP_001399.1 2
XM_005270580.4 XP_005270637.1 2