gRNA Design Details

Target DNA Sequence
CGTTCACTACCAGCATGGCA
PAM Sequence
CGG
Target locus
Chr.3: 114147514 - 114147536 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 92.44/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
DRD3
Entrez Gene ID
1814
Gene Name
dopamine receptor D3
Gene Aliases
D3DR, ETM1, FET1
Chromosome Location
Chr.3: 114127623 - 114199407 on GRCh38
UniGene ID
Hs.121478
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000796.5 NP_000787.2 4
NM_001282563.2 NP_001269492.1 5
NM_001290809.1 NP_001277738.1 5
NM_033663.5 NP_387512.3 4
XM_017005829.1 XP_016861318.1 5