gRNA Design Details
- Target DNA Sequence
- GGTTATCGTCTCTGTGCGAG
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 236178513 - 236178535 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
90.84/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GPR137B
- Entrez Gene ID
-
7107
- Gene Name
- G protein-coupled receptor 137B
- Gene Aliases
-
TM7SF1
- Chromosome Location
-
Chr.1: 236142502 - 236211059 on GRCh38
- UniGene ID
- Hs.498160
- Species
-
Human