gRNA Design Details

Target DNA Sequence
GATTCACAACCGAATTGGTG
PAM Sequence
TGG
Target locus
Chr.22: 24441081 - 24441103 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 93.54/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ADORA2A
Entrez Gene ID
135
Gene Name
adenosine A2a receptor
Gene Aliases
A2aR, ADORA2, RDC8
Chromosome Location
Chr.22: 24423597 - 24442360 on GRCh38
UniGene ID
Hs.197029
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000675.5 NP_000666.2 3
NM_001278497.1 NP_001265426.1 4
NM_001278498.1 NP_001265427.1 3
NM_001278499.1 NP_001265428.1 3
NM_001278500.1 NP_001265429.1 3
NR_103543.1 2
NR_103544.1 2