gRNA Design Details
- Target DNA Sequence
- CTCTGCACGGCTCGCCTGGT
- PAM Sequence
- CGG
- Target locus
-
Chr.1: 6555380 - 6555402 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
93.73/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TAS1R1
- Entrez Gene ID
-
80835
- Gene Name
- taste 1 receptor member 1
- Gene Aliases
-
GM148, GPR70, T1R1, TR1
- Chromosome Location
-
Chr.1: 6555278 - 6579757 on GRCh38
- UniGene ID
- Hs.124574
- Species
-
Human