gRNA Design Details
- Target DNA Sequence
- TTTGCCACAAAGGAGCGCGA
- PAM Sequence
- CGG
- Target locus
-
Chr.1: 109261514 - 109261536 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
92.92/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CELSR2
- Entrez Gene ID
-
1952
- Gene Name
- cadherin EGF LAG seven-pass G-type receptor 2
- Gene Aliases
-
ADGRC2, CDHF10, EGFL2, Flamingo1, MEGF3
- Chromosome Location
-
Chr.1: 109249705 - 109275756 on GRCh38
- UniGene ID
- Hs.57652
- Species
-
Human