gRNA Design Details

Target DNA Sequence
TGACGCGGAACTTCTGGATG
PAM Sequence
CGG
Target locus
Chr.1: 203165789 - 203165811 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 96.73/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ADORA1
Entrez Gene ID
134
Gene Name
adenosine A1 receptor
Gene Aliases
RDC7
Chromosome Location
Chr.1: 203127705 - 203167405 on GRCh38
UniGene ID
Hs.77867
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000674.2 NP_000665.1 4
NM_001048230.1 NP_001041695.1 3
XM_005244901.1 XP_005244958.1 4
XM_005244902.4 XP_005244959.1 2
XM_017000342.1 XP_016855831.1 3