gRNA Design Details
- Target DNA Sequence
- TGGTTTGGCATTTCCTTCCG
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 23379026 - 23379048 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
73.98/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ogfrl1
- Entrez Gene ID
-
70155
- Gene Name
- opioid growth factor receptor-like 1
- Gene Aliases
-
2210417C17Rik
- Chromosome Location
-
Chr.1: 23366424 - 23383210 on GRCm38
- UniGene ID
- Mm.28013
- Species
-
Mouse