gRNA Design Details
- Target DNA Sequence
- AGCGGCTACTCTTAGGTGTC
- PAM Sequence
- TGG
- Target locus
-
Chr.7: 27335662 - 27335684 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85.73/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ltbp4
- Entrez Gene ID
-
108075
- Gene Name
- latent transforming growth factor beta binding protein 4
- Gene Aliases
-
2310046A13Rik
- Chromosome Location
-
Chr.7: 27305140 - 27339709 on GRCm38
- UniGene ID
- Mm.272251
- Species
-
Mouse