gRNA Design Details
- Target DNA Sequence
- ATTGCTAGAGTTCACCCAAG
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 121605707 - 121605729 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
71.68/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CLASP1
- Entrez Gene ID
-
23332
- Gene Name
- cytoplasmic linker associated protein 1
- Gene Aliases
-
MAST1
- Chromosome Location
-
Chr.2: 121337776 - 121649476 on GRCh38
- UniGene ID
- Hs.469840
- Species
-
Human