gRNA Design Details
- Target DNA Sequence
- AGATTACCGCTATGGGCGAG
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 137519158 - 137519180 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
91.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gigyf1
- Entrez Gene ID
-
57330
- Gene Name
- GRB10 interacting GYF protein 1
- Gene Aliases
-
Perq1, mKIAA4110
- Chromosome Location
-
Chr.5: 137510258 - 137527935 on GRCm38
- UniGene ID
- Mm.268344
- Species
-
Mouse