gRNA Design Details
- Target DNA Sequence
- ATGCTTGGTAGACCCCCAGC
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 45615219 - 45615241 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
82.95/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fgf21
- Entrez Gene ID
-
56636
- Gene Name
- fibroblast growth factor 21
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 45613890 - 45619350 on GRCm38
- UniGene ID
- Mm.143736
- Species
-
Mouse