gRNA Design Details
- Target DNA Sequence
- ACATGGTTTTCTCCCCGTGG
- PAM Sequence
- CGG
- Target locus
-
Chr.18: 57191128 - 57191150 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.55/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Megf10
- Entrez Gene ID
-
70417
- Gene Name
- multiple EGF-like-domains 10
- Gene Aliases
-
3000002B06Rik, Gm331
- Chromosome Location
-
Chr.18: 57133090 - 57297467 on GRCm38
- UniGene ID
- Mm.297863
- Species
-
Mouse