gRNA Design Details
- Target DNA Sequence
- GAACAAAACCCTCCGCTCCT
- PAM Sequence
- CGG
- Target locus
-
Chr.11: 61660266 - 61660288 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.13/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Grap
- Entrez Gene ID
-
71520
- Gene Name
- GRB2-related adaptor protein
- Gene Aliases
-
8430435N19Rik, BB233594
- Chromosome Location
-
Chr.11: 61653321 - 61672777 on GRCm38
- UniGene ID
- Mm.202760
- Species
-
Mouse