gRNA Design Details
- Target DNA Sequence
- GAGCCGTGGTGGTGTTTGTG
- PAM Sequence
- AGG
- Target locus
-
Chr.13: 64312493 - 64312515 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
77.77/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Aaed1
- Entrez Gene ID
-
66129
- Gene Name
- AhpC/TSA antioxidant enzyme domain containing 1
- Gene Aliases
-
1110018J18Rik, AI849003
- Chromosome Location
-
Chr.13: 64291836 - 64312710 on GRCm38
- UniGene ID
- Mm.491110
- Species
-
Mouse