gRNA Design Details
- Target DNA Sequence
- GCCCATCGCTTGCATCATGA
- PAM Sequence
- TGG
- Target locus
-
Chr.8: 40938231 - 40938253 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.55/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Pdgfrl
- Entrez Gene ID
-
68797
- Gene Name
- platelet-derived growth factor receptor-like
- Gene Aliases
-
1110039P19Rik, AV013190
- Chromosome Location
-
Chr.8: 40926233 - 40990782 on GRCm38
- UniGene ID
- Mm.284246
- Species
-
Mouse