gRNA Design Details
- Target DNA Sequence
- GCACGAACTTGAGCTCGTTG
- PAM Sequence
- AGG
- Target locus
-
Chr.4: 58206055 - 58206077 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
91.07/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Svep1
- Entrez Gene ID
-
64817
- Gene Name
- sushi, von Willebrand factor type A, EGF and pentraxin domain containing 1
- Gene Aliases
-
1110021D17Rik, 4833413O10Rik, AA387071, D430029O09Rik, Polydom
- Chromosome Location
-
Chr.4: 58042796 - 58206596 on GRCm38
- UniGene ID
- Mm.261024
- Species
-
Mouse