gRNA Design Details
- Target DNA Sequence
- CAATGCCATTCTCCCGCAGG
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 131273787 - 131273809 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
90.22/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ikbke
- Entrez Gene ID
-
56489
- Gene Name
- inhibitor of kappaB kinase epsilon
- Gene Aliases
-
AW558201, IKK-i, IKKepsilon, Ikki
- Chromosome Location
-
Chr.1: 131254600 - 131279625 on GRCm38
- UniGene ID
- Mm.386783
- Species
-
Mouse