gRNA Design Details
- Target DNA Sequence
- CGAAGAGACTGTGATCCTGC
- PAM Sequence
- CGG
- Target locus
-
Chr.8: 40286422 - 40286444 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85.07/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fgf20
- Entrez Gene ID
-
80857
- Gene Name
- fibroblast growth factor 20
- Gene Aliases
-
-
- Chromosome Location
-
Chr.8: 40279166 - 40286953 on GRCm38
- UniGene ID
- Mm.425526
- Species
-
Mouse