gRNA Design Details
- Target DNA Sequence
- CAGCGCTTTTTCAGCGGCTA
- PAM Sequence
- CGG
- Target locus
-
Chr.2: 162931725 - 162931747 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
90.18/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Srsf6
- Entrez Gene ID
-
67996
- Gene Name
- serine/arginine-rich splicing factor 6
- Gene Aliases
-
1210001E11Rik, AI314910, AW146126, Sfrs6
- Chromosome Location
-
Chr.2: 162931508 - 162937121 on GRCm38
- UniGene ID
- Mm.24042
- Species
-
Mouse