gRNA Design Details
- Target DNA Sequence
- GTACTACTTCACCAGGAGGC
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 66002370 - 66002392 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
89.23/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Csrnp3
- Entrez Gene ID
-
77771
- Gene Name
- cysteine-serine-rich nuclear protein 3
- Gene Aliases
-
A330102K23Rik, CSRNP-3, Csnrp3, Mbu1, taip-2
- Chromosome Location
-
Chr.2: 65845767 - 66031546 on GRCm38
- UniGene ID
- Mm.102025
- Species
-
Mouse