gRNA Design Details
- Target DNA Sequence
- CAGCAACGGGATCAAGATGT
- PAM Sequence
- AGG
- Target locus
-
Chr.19: 10634502 - 10634524 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.73/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Vwce
- Entrez Gene ID
-
71768
- Gene Name
- von Willebrand factor C and EGF domains
- Gene Aliases
-
1300015B04Rik
- Chromosome Location
-
Chr.19: 10634229 - 10665210 on GRCm38
- UniGene ID
- Mm.169261
- Species
-
Mouse