gRNA Design Details
- Target DNA Sequence
- GCAGTTCTCCTCGACGAGAG
- PAM Sequence
- AGG
- Target locus
-
Chr.17: 23810424 - 23810446 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.51/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Srrm2
- Entrez Gene ID
-
75956
- Gene Name
- serine/arginine repetitive matrix 2
- Gene Aliases
-
5033413A03Rik, AA410130, SRm300, mKIAA0324
- Chromosome Location
-
Chr.17: 23790857 - 23824741 on GRCm38
- UniGene ID
- Mm.436876
- Species
-
Mouse