gRNA Design Details
- Target DNA Sequence
- AGTGACCACCATTTCTGCGG
- PAM Sequence
- CGG
- Target locus
-
Chr.14: 64081722 - 64081744 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.92/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prss55
- Entrez Gene ID
-
71037
- Gene Name
- protease, serine 55
- Gene Aliases
-
4933401F05Rik
- Chromosome Location
-
Chr.14: 64075435 - 64085433 on GRCm38
- UniGene ID
- Mm.19977
- Species
-
Mouse