gRNA Design Details
- Target DNA Sequence
- GCCTTTGTGCCGGATTATGG
- PAM Sequence
- TGG
- Target locus
-
Chr.14: 64079353 - 64079375 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.19/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prss55
- Entrez Gene ID
-
71037
- Gene Name
- protease, serine 55
- Gene Aliases
-
4933401F05Rik
- Chromosome Location
-
Chr.14: 64075435 - 64085433 on GRCm38
- UniGene ID
- Mm.19977
- Species
-
Mouse