gRNA Design Details
- Target DNA Sequence
- GTGGAACGGGAACTGCTGAT
- PAM Sequence
- GGG
- Target locus
-
Chr.1: 43075880 - 43075902 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86.4/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tgfbrap1
- Entrez Gene ID
-
73122
- Gene Name
- transforming growth factor, beta receptor associated protein 1
- Gene Aliases
-
3110018K12Rik, AU024090, AU042859, AW742551, Trap1
- Chromosome Location
-
Chr.1: 43047265 - 43098622 on GRCm38
- UniGene ID
- Mm.246069
- Species
-
Mouse