gRNA Design Details
- Target DNA Sequence
- GCGTGTACAGTGGATGCCCA
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 87183552 - 87183574 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.02/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prss56
- Entrez Gene ID
-
69453
- Gene Name
- protease, serine 56
- Gene Aliases
-
1700027L20Rik
- Chromosome Location
-
Chr.1: 87183094 - 87188405 on GRCm38
- UniGene ID
- Mm.84618
- Species
-
Mouse